Learn More
ACROBiosystems Human FOLR1 25tests
Supplier:  ACROBiosystems FO1HP2H625TESTS
Encompass_Preferred
PE-Labeled Human FOLR1 Protein His Tag (FO1-HP2H6) is produced via conjugation of PE to Human FOLR1 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human FOLR1 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).
Catalog No. 50-253-5051
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.