Learn More
ACROBiosystems Mouse CXCR3 20ug
Supplier:  ACROBiosystems CX3M52P720UG
Encompass_Preferred
Mouse CXCR3 Full Length Protein (VLP) (CX3-M52P7) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 - Leu 367 (Accession # O88410).
Catalog No. 50-253-5188
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.