Learn More
ACROBiosystems Human CD19 2Vials
Supplier:  ACROBiosystems CHEKATP0032VIALS
Encompass_Preferred
Chimeric antigen receptors (CARs) comprise interact domains of which target cancer cell expressed antigens and allow T cell to efficiently kill the cancer cell. FMC63 is an IgG2a mouse monoclonal antibody specific for CD19 which is a target for the immunotherapy of B lineage leukaemias and lymphomas. FMC63 scFv is the most commonly used ectodomain component of CD19-specific CARs. Our CAR-like cells apply the HEK293 cells for the expression of FMC63 which will be presented on the cell membrane. It mimics the real CAR-T cell reaction and it is used as positive control for Anti-CD19 scFv Antibody flow cytometry detection.
Catalog No. 50-253-5291
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.