Learn More
ACROBiosystems Human CD27 Ligand 200ug
Supplier:  ACROBiosystems CDLHA245200UG
Encompass_Preferred
Alexa Fluor 555-Labeled Human CD27 Ligand Protein His Tag (CDL-HA245) is produced via conjugation of AF555 to Human CD27 Ligand Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD27 Ligand Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1).
Catalog No. 50-253-5309
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.