Learn More
ACROBiosystems Cynomolgus CD39 20ug
Supplier:  ACROBiosystems CD9C5PH320UG
Encompass_Preferred
Cynomolgus CD39 Protein His Tag (CD9-C5PH3) is expressed from CHO cells. It contains AA Thr 60 - Val 500 (Accession # XP_015311944.1).
Catalog No. 50-253-5332
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.