Learn More
ACROBiosystems Mouse CD206 200ug
Supplier:  ACROBiosystems CD6M82E8200UG
Encompass_Preferred
Biotinylated Mouse CD206 Protein HisAvitag (CD6-M82E8) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Ala 1388 (Accession # Q61830).
Catalog No. 50-253-5355
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.