Learn More
ACROBiosystems Human APOE3 Protein Tag Free
Supplier:  ACROBiosystems APEH52161MG
Encompass_Preferred
Human APOE3 Protein Tag Free (APE-H5216) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession # P02649-1 ).
Catalog No. 50-253-5498
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.