Learn More
World Precision Instrument MRI-Compatible Ventilator for small animals
Supplier: World Precision Instrument CWMRI1
The CWE-MRI-1 Ventilator is a small animal ventilator designed for use in MRI and other high magnetic field environments, though its use is not limited to those environments. Operated alone or controlled/monitored by a computer, the CWE-MRI-1 system consists of a microprocessor-based control unit and a set of remote, pneumatically operated, nonmetallic valves. The CWE-MRI-1 operates on the flow-time principle, dispensing a known airflow into the lungs for a set inspiratory time to generate the desired tidal volumes. The three controls – respiratory rate, inspiratory time and flow rate – allow for accuracy and extraordinary flexibility over a wide range of volumes, breaths-per-minute and inspiration/expiration time (I/E) ratios. An internal air pump generates the respiratory airflow, but CWE-MRI-1can also connect to an external pressurized air or gas source (oxygen or anesthetic gasses). The remote high-speed, miniature pneumatic valves control the airflow switching.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.