Learn More
ORS Nasco ULTRSPC 2001 DUO TEM CL XTR1PR
Supplier: ORS Nasco 763S0112C
Honeywell part# S0112C. Honeywell Uvex Ultraspec 2001 OTG Safety Glasses Lens Type: Anti-fog Lens Tint: Clear Lens Coating: Uvextreme Anti-fog UV Protection: 99.90% Certifications/Compliance: ANSI Z87.1-2015 CSA Z94.3-2010 Eyewear Recommended Applications: Safety Temple Style: Duoflex Weight: 51g
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.