Learn More
Amsbio LLC Ab Heparan Sulfate, BIOTINYLATED (HIGH BIOTIN) clone F58-10E4, 50ug pack
Supplier:  Amsbio LLC 370255B50H
Encompass_Preferred
Biotinylated antibodies offer advantages in several key applications, by cutting out the need for the use of conjugated secondary antibodies to quantify the level of detection/binding in immunological assays. The antibody (F58-10E4 clone) reacts with the 10E4 epitope, which is present in many types of heparan sulfate. 10E4 epitope includes (an) N-sulfated glucosamine residue(s) that are critical for the reactivity of the F58-10E4 antibody. The reactivity of the antibody with most heparan sulfates is nearly completely abolished after treatment of the glycosaminoglycan with bacterial heparatinase I (Heparinase III) from Flavobacterium heparinum (EC 4.2.2.8). The 10E4 antibody does not react with hyaluronan, chondroitin sulfate, dermatan sulfate, keratan sulfate or DNA. Host species: Mouse, Isotype: IgM, Label/Conjugate: Biotin, Species reactivity: Broad, Storage: 2 - 8 deg C.
Catalog No. 50-256-6539
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.