Learn More
ACROBiosystems resDetect™ Human FGF2 ELISA Kit (Residue Testing)
Supplier:  ACROBiosystems RESA00996TESTS
Encompass_Preferred
Human FGF2 ELISA Kit is based on ELISA sandwich method and designed to measure human FGF2 levels in cell culture supernates, serum, and plasma. It contains recombinant human FGF2 and a pair of antibodies against the recombinant human FGF2, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP human FGF2 (ACROBiosystems, cat#GMP-FGCH17) concentrations. The specificity has been verified.
Catalog No. 50-271-8982
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.