Learn More
ACROBiosystems Human IL-3 Protein, premium grade
Supplier:  ACROBiosystems IL3H51151MG
Encompass_Preferred
Interleukin-3 (IL-3) is an interleukin, a type of biological signal (cytokine) which is encoded by the IL-3 gene located on chromosome 5 and produced primarily by activated T cells beside human thymic epithelial cells, activated murine mast cells, murine keratinocytes and neurons/astrocytes. The protein acts in hematopoiesis by controlling the production, differentiation, and function of 2 related white cell populations of the blood, the granulocytes and the monocytes-macrophages. The human IL-3 reported to be a monomer, as it is known, contains 133 amino acids residues which is a single non-glycosylated polypeptide. Specifically, human and murine IL-3 share low homology and it does not show activity on murine cells.
Catalog No. 50-271-9340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.