Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01&B2M&EBV LMP1 (YLLEMLWRL) Complex Protein (Monomer, MALS verified)
Supplier:  ACROBiosystems HLPH82EC25UG
Encompass_Preferred
Epstein-Bar Virus (EBV), also known as human herpesvirus 4, belongs to gamma herpes virus family and is a very common human virus worldwide. EBV causes infectious mononucleosis (IM) and also associates to some specific types of cancers such as Burkitt’s lymphoma (BL) and gastric carcinoma (GC). Glycoprotein B (gB) plays an important role in viral entry by binding with αvβ6/αvβ8 integrins to trigger the membrane fusion and entry process of epithelial cells, which makes it become an great target for EBV research. Epstein-Bar Virus (EBV), also known as human herpesvirus 4, belongs to gamma herpes virus family and is a very common human virus worldwide. EBV causes infectious mononucleosis (IM) and also associates to some specific types of cancers such as Burkitt’s lymphoma (BL) and gastric carcinoma (GC). Glycoprotein B (gB) plays an important role in viral entry by binding with αvβ6/αvβ8 integrins to trigger the membrane fusion and entry process of epithelial cells, which makes it become
Catalog No. 50-271-9410
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.