Learn More
ACROBiosystems Biotinylated Human HLA-B*15:02&B2M Monomer Protein (Peptide free, MALS verified)
Supplier:  ACROBiosystems HLMH82EQ25UG
Encompass_Preferred
Biotinylated Human HLA-B*15:02&B2M Monomer Protein (HLM-H82Eq) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*15:02) & Ile 21 - Met 119 (B2M) (Accession # AAA59630.1 (HLA-B*15:02) & P61769 (B2M)).
Catalog No. 50-271-9438
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.