Learn More
ACROBiosystems ELISA 20× Washing Buffer
Supplier:  ACROBiosystems CEBC30150ML
Encompass_Preferred
ELISA Washing Buffer is a PBS-based wash buffer for use in sandwich ELISA procedures. The buffer is the same formulation that is supplied with most ClinMax™ ELISA kits for cytokines and biomarkers, but not all. The 20X concentrated stock solution provided should be diluted to create a 1X working buffer for use in ELISA.
Catalog No. 50-271-9758
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.