Learn More
ACROBiosystems Cynomolgus CDCP1 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems CD1C52H3100UG
Encompass_Preferred
CUB domain-containing protein 1, also know as CDCP1. CDCP1 is a cell surface glycoprotein that has been recognized both as a tumour marker and as a potential target to disrupt progression of cancer. CDCP1 has been shown to modulate cellsubstratum adhesion and motility in colon cancer cell lines.
Catalog No. 50-271-9845
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.