Learn More
PHCbi Cell-IQ™ Series 5.8 cu.ft. CO2 Laboratory Incubato
Supplier: PHCbi MCO170AICUVLPA
The PHCbi Cell-IQ CO2 incubator design and touchscreen panel deliver exceptional ease of use, effortless cleaning and maintenance as well as outstanding performance with multilevel contamination control.The MCO-170AICUVL-PA laboratory incubator is designed for demanding and regulated applications in the biomedical, pharmaceutical, medical research and clinical laboratory applications. The Cell-IQ CO2 incubator offers the ideal environment for all your research needs. The MCO-170AICUVL-PA unit includes a touchscreen control panel that delivers improved usability, an optimized interior structure with technology for rapid cleaning, and a proprietary infrared sensor for stabilizing temperature, CO2, and humidity levels.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.