Learn More
New England Biolabs, Inc. Monarch Spin Plasmid Miniprep Kit - 50 preps
Supplier: New England Biolabs, Inc. T1110S
The Monarch Spin Plasmid Miniprep kit, a newer version of our original Monarch Miniprep kit, offers a unique method for plasmid preparation and purification, enabling extraction of highly concentrated plasmid DNA from various bacterial strains, with yields of up to 20 ug of high-quality DNA. This kit features upgraded spin columns precision-engineered to uniquely allow for low elution, in as little as 30 ul, for highly concentrated DNA in downstream application such as sequencing, restriction digestion, transformation, transfection, and more. A colored buffer system helps indicate completion of key lysis and neutralization steps for an easy to follow workflow and shorter, faster protocols allow for reduced hands-on time and less spin time. Compared to leading suppliers, these kits use significantly less plastic through uniquely designed columns and bottles with plastic reduction while still maintaining structural integrity for centrifugation or vacuum manifolds and other steps
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.