Learn More
Cayman Chemical Glycogen HydrolysIs BuFR 10mL
Supplier: Cayman Chemical 70048410 ML
A GPR52 agonist increases cAMP concentration in CHO cells expressing human GPR52 (EC50 75 nM) selective for GPR52 over 98 other receptors ion channels and enzymes at 10 UM increases c-Fos expression in the nucleus accumbens shell to a greater extent than in the dorsomedial striatum at 30 mg/kg increases the time spent with the novel object in the novel object recognition test in rats at 3 and 10 mg/kg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.