Learn More
Sigma Aldrich Fine Chemicals Biosciences Zone-freeTM Sealing films
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z72164650EA
Encompass
With a 70 mum polyethylene top layer and inert white polypropylene and acrylic adhesive sublayer this easy-piercing sealing film allows direct sample recovery with single or multichannel pipettors and robotic probes. The adhesive-free zone above each well prevents adhesive fouling of the pipet tip or probe ultimately minimizing the amount of adhesive that comes into contact with the sample. Dimensions are 79.4 by 142.9 mm for 96-well plates.
Catalog No. 50-290-1023
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.