Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum

Catalog No. 503100162 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
10 Ea.
5 Ea.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-310-0162 10 Ea.
50-309-8083 5 Ea.
50-310-0167 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-310-0162 Supplier Sigma Aldrich Supplier No. GF5915056110EA
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Molybdenum

Specifications

CAS 7439-98-7
Melting Point 2617°C
Boiling Point 4612°C
Density 10.3 g/mL (at 25°C)
Quantity 10 Ea.
Molecular Formula Mo
Linear Formula Mo
MDL Number MFCD00003465
Synonym Molybdenum; MO000203
Molecular Weight (g/mol) 95.94
Percent Purity 99.9%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.