Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lead

Catalog No. 503120580 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
10 Ea.
5 Ea.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-0580 10 Ea.
50-312-2192 1 Ea.
50-312-2027 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-0580 Supplier Sigma Aldrich Supplier No. GF7425979310EA
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lead

Specifications

Melting Point 327.4°C
CAS 7439-92-1
Boiling Point 1740°C
Quantity 10 Ea.
Molecular Formula Pb
Linear Formula Pb
MDL Number MFCD00134050
Synonym Lead; PB000413
Molecular Weight (g/mol) 207.2
Percent Purity 99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.