Learn More
Sigma Organic Chemistry Methyl acrylate | 250ML | 96-33-3 | MFCD00008627 | 0.99
Supplier: Sigma Organic Chemistry M27301250ML
Methyl acrylate, 250ML
About This Item:
Linear Formula: CH2=CHCOOCH3
Molecular Weight: 86.09
CAS Number: 96-33-3
MDL Number: MFCD00008627
UNSPSC Code: 12162002
Purity: 0.99
Boiling Point: 80 C (lit.)
Melting Point: -75 C (lit.)
Density: 0.956 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.402 (lit.)
Storage: 2-8C
EINECS Number: 202-500-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.