Learn More
Sigma Organic Chemistry Styrene oxide | 5G | 96-09-3 | MFCD00005121 | 0.97
Supplier: Sigma Organic Chemistry S50065GA

Styrene oxide, 5G
About This Item:
Synonym(s): 1,2-Epoxyethylbenzene; Phenylethylene oxide; Phenyloxirane
Linear Formula: C8H8O
Molecular Weight: 120.15
CAS Number: 96-09-3
MDL Number: MFCD00005121
UNSPSC Code: 12162002
Purity: 0.97
Boiling Point: 194 C (lit.)
Melting Point: -37 C (lit.)
Density: 1.054 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.535 (lit.)
Storage: room temp
EINECS Number: 202-476-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.