Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicone Oil Ap 150 Wacker

Catalog No. 50384250MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
Catalog No. Quantity
50-384-250MLF 250 mL
50-384-1LF 1 L
2 options

Catalog No. 50-384-250MLF

Supplier: Sigma Aldrich 50384250MLF

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil AP 150 Wacker is a conventional electrorheologic (ER) fluid. It was employed as an insulating medium during the poling of hard ferroelectric ceramics. Silicon oil is composed of a mixture of hydrophobic polymeric and monomeric compounds, containing silicon-oxygen bonds. Its physical and chemical characteristics have been reported. The utility of silicon oil as a vitreous substitute has been studied.Silicone oil AP 150 Wacker was used to prevent the water evaporation during rheological characterization of injectable polysaccharide system based on calcium alginate hydrogel and two dextran methacrylate derivatives. It may be used for the determination of chloroplast volume by the silicone oil filtering centrifugation technique.

Specifications

CAS 63148-52-7
Refractive Index n20/D 1.51
MDL Number MFCD05865496
Quantity 250 mL
Synonym Wacker Silicon oil AP 150
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.