Learn More
Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits
Shop All Spectrum Medical Industries Products

Description
Includes
- 1m or 5m of Spectra/Por 2 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
- For higher permeability applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water)
Specifications
Specifications
| MWCO | 12 to 14 kDa |
| Diameter (Metric) | 67 mm |
| Length (Metric) | 34 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.