Learn More
Cellpath System II Hex™ Tissue Processing Cassettes
SDP
Catalog No. 22222064
Change view
Click to view available options
Color:
Blue
Brown
Dark Blue
Dark Green
Gray
Green
Lavender
Orange
Peach
Pink
Red
Tan
Turquoise
Yellow
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-222-064 Pink
22-222-068 Tan
22-222-067 Peach
22-222-066 Red
22-222-065 Yellow
22-222-073 Turquoise
22-222-072 Lavender
22-222-071 Dark Green
22-222-070 Dark Blue
22-222-069 Brown
22-222-063 Orange
22-222-062 Blue
22-222-060 Green
22-222-061 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 22-222-064 Supplier Cellpath Supplier No. EAD010803A
This item is not returnable. View return policy
To hold tissue specimens during embedding, processing, and storage. CellPath System II Hex Tissue Processing Cassettes feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
  • Manufactured from high–grade virgin acetyl resin
  • Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
  • Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
  • Extra thick side walls increases rigidity and stability for high quality sectioning
  • Available in 15 stock colors which color code different types of workload or applications
Specifications
Quantity 1000/Pk.
Color Pink
Material Acetyl Resin
Product Type Tissue Processing Cassette