Learn More
Cellpath System II Hex™ Tissue Processing Cassettes (Hoppered)
SDP
Catalog No. 22222081
Change view
Click to view available options
Color:
Blue
Brown
Dark Blue
Dark Green
Gray
Green
Lavender
Peach
Pink
Red
Turquoise
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-222-081 Red
22-222-088 Turquoise
22-222-086 Dark Green
22-222-087 Lavender
22-222-085 Dark Blue
22-222-084 Brown
22-222-080 Yellow
22-222-082 Peach
22-222-079 Pink
22-222-077 Blue
22-222-075 Green
22-222-076 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 22-222-081 Supplier Cellpath Supplier No. EAD011004H
Item Discontinued This product is either new or inactive, and cannot be ordered online.
Pre-loaded into hoppers for use with automated and carousel type printers. CellPath System II Hex Tissue Processing Cassettes (Hoppered) feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
  • Pre-loaded into hoppers for use with automated and carousel type cassette printers
  • Manufactured from high–grade virgin acetyl resin
  • Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
  • Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
  • Extra thick side walls increases rigidity and stability for high quality sectioning
  • Designed to suit Thermo Printmate and Micro-writer cassette–writing machines
  • Available in 15 stock colors which color code different types of workload or applications
Specifications
Quantity 1500/Pk.
Color Red
Material Acetyl Resin
Product Type Tissue Processing Cassette in Hopper tube