Learn More
Cellpath System II Hex™ Tissue Processing Cassettes (Hoppered)
Description
- Pre-loaded into hoppers for use with automated and carousel type cassette printers
- Manufactured from high–grade virgin acetyl resin
- Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
- Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
- Extra thick side walls increases rigidity and stability for high quality sectioning
- Designed to suit Thermo Printmate and Micro-writer cassette–writing machines
- Available in 15 stock colors which color code different types of workload or applications
Specifications
Specifications
| Quantity | 1500/Pk. |
| Color | Brown |
| Material | Acetyl Resin |
| Product Type | Tissue Processing Cassette in Hopper tube |