Learn More
Cellpath Stainless-steel Reusable Base Molds
Description
- Made of reusable Stainless steel
- Stackable to reduce storage area
- Internal rounded corners enable easy removal of the wax block
- Features extended edges prevent tipping and provide grip area for ease of handling
- Available in wide range of sizes
- GBB-0714-05A suitable for use with needle and punch biopsies
Specifications
Specifications
| Material | Stainless Steel |
| Dimensions (L x W x H) | 10 x 24 x 5 mm |
| Quantity | 5/Pk. |