Learn More
Sino Biological Mouse Ephrin-A2 / EFNA2 Protein 200ug
Supplier:  Sino Biological 50579MCCH200
Encompass_Preferred
A DNA sequence encoding the mouse EFNA2 (NP_031935.3) without the pro peptide (Met 1-Asn 184) was expressed and purified, with additional seven amino acid (ALEVLFQ) at the C-terminus.
Catalog No. 50-162-2139
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.