Learn More
Sino Biological Mouse LAMP2 / CD107b Protein (His Tag) 100ug
Supplier:  Sino Biological 50791M08H100
Encompass_Preferred
A DNA sequence encoding the mouse LAMP2 (P17047-1) extracellular domain (Leu 26-Asn 379) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-162-2502
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.