Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

508.1 Pesticide Kit, Restek™
SDP

Catalog No. 06719745 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
7 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-745 7 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-745 Supplier Restek Corporation Supplier No. 32097
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Compounds:

32045: 508 Performance Check Mix

δ-BHC (319-86-8), 0.4 μg/mL
Chlorothalonil (1897-45-6), 0.5 μg/mL
Chlorpyrifos (2921-88-2), 0.02 μg/mL
DCPA methyl ester (Chlorthal-dimethyl) (1861-32-1), 0.5 μg/mL

32091: 508.1 Internal Standard
Pentachloronitrobenzene (82-68-8)

32092: 508.1 Surrogate
4,4'-Dibromobiphenyl (92-86-4)

32093: 508.1 GC Degradation Check Mix
4,4'-DDT (50-29-3)
Endrin (72-20-8)

32094: 508.1 Calibration Mix #1
Aldrin (309-00-2)
α-BHC (319-84-6)
β-BHC (319-85-7)
δ-BHC (319-86-8)
γ-BHC (Lindane) (58-89-9)
4,4'-DDD (72-54-8)
4,4'-DDE (72-55-9)
4,4'-DDT (50-29-3)
Dieldrin (60-57-1)
Endosulfan I (959-98-8)
Endosulfan II (33213-65-9)
Endosulfan sulfate (1031-07-8)
Endrin (72-20-8)
Endrin aldehyde (7421-93-4)
Heptachlor (76-44-8)
Heptachlor epoxide (isomer B) (1024-57-3)
Methoxychlor (72-43-5)

32095: 508.1 Calibration Mix #2
Chlorobenzilate (510-15-6)
cis-Chlordane (5103-71-9)
trans-Chlordane (5103-74-2)
Chloroneb (2675-77-6)
DCPA methyl ester (Chlorthal-dimethyl) (1861-32-1)
Etridiazole (2593-15-9)
Hexachlorobenzene (118-74-1)
cis-Permethrin* (52645-53-1)
trans-Permethrin* (52645-53-1)
Propachlor (1918-16-7)
Trifluralin (1582-09-8)

32096: 508.1 Calibration Mix #3
Alachlor (15972-60-8)
Atrazine (1912-24-9)
Chlorothalonil (1897-45-6)
Cyanazine (bladex) (21725-46-2)
Hexachlorocyclopentadiene (77-47-4)
Metolachlor (51218-45-2)
Metribuzin (21087-64-9)
Simazine (122-34-9)

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

CAS Max % 99.999858
Odor Moderate, Ester-like
Vapor Density 3.04 (Air = 1)
Specific Gravity 0.74g/cm3 at 20°C
Concentration Contains 1mL each of these mixtures.
UN Number UN1173
Melting Point -108.6°C
Type Multi-component Standard
Quantity 7 x 1 mL
Chemical Name or Material Methyl-tert-butyl ether
ShelfLife 6 months
EINECSNumber 216-653-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.