Learn More
Sino Biological Mouse NGFR / P75 Protein (His Tag) 100ug
Supplier:  Sino Biological 50971M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse NGFR (Q9Z0W1) (Met 1-Asn 243), without the propeptide, was fused with a C-terminal polyhistidine tag.
Catalog No. 50-162-2702
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.