Learn More
Sino Biological Mouse S100B / S100beta Protein (Fc Tag) 100ug
Supplier:  Sino Biological 50986M01H100
Encompass_Preferred
A DNA sequence encoding the mouse S100B (P50114) (Ser2-Glu92) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-2722
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.