Learn More
Bellco Glass, Inc. 50mL Pipet Bulk Packed
SDP
Supplier:  Bellco Glass, Inc. 229250000
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bidirectional positive and negative graduations are printed in bold black ink 10mL wide tip Serological pipet for viscous solutions. Wide tip is 4x larger than standard 10mL pipet 25 amp 50mL pipet features an antidrip tip for accurate delivery Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Color Code Purple 30 Individually Wrapped Case Quantity 90
Catalog No. 50-232-9955
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.