Learn More
Biopioneer 50 mL Serological Pipet, 100/pk
SDP
Supplier:  Biopioneer GEX500S01100PK
Encompass_Preferred
50ml Serological Pipet, Individual Wrapped (one side paper, one side plastic), Sterilized pipet can be used for variety of purpose, 100/pk. BioPioneer Serological pipettes are sterilized by Gamma irradiation, certified Non-pyrogenic, DNase/RNase Free.
Catalog No. NC1891001