Learn More
Nanocomposix Ultra Uniform Gold Nanoparticle Number Standard - 50 nm, 1E8 particles/mL, 25 mL
SDP
Supplier:  Nanocomposix AUXS5025M
Encompass_Preferred
Ultra Uniform materials are our most highly-monodispersed particles. Ideal for use as size standards, applications requiring a tight size distribution. Our Ultra Uniform Particle Number Standards provide additional characterization and careful formulation for applications that require a known particle number concentration.PEG-carboxyl is covalently bound to the metal surface. The acid provides a highly negatively charged surface and can be utilized for conjugation.Provided at 10^8 particles/mL in aqueous 1 mM sodium citrate.APPLICATIONS- sp-ICP-MS: Single Particle Inductively Coupled Plasma Mass Spectroscopy- NTA: Nano Tracking Analysis- TRPS: Tunable Resistive Pulse Sensing
Catalog No. NC2885887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.