Learn More
JDM SCIENTIFIC LLC The LabBee Checkit Go - 50UL
Supplier:  JDM SCIENTIFIC LLC MSLO8C506
Encompass_Preferred
The LabBee Checkit Go is a simple, stand-alone, disposable cartridge that allows you to check the accuracy of your liquid handling robot or multi-channel pipette in just seconds. No additional equipment is required.
The easy to use LabBee branded Checkit Go device measures the dispensed volumes of up to 8 channels simultaneously in less than 10 seconds. The cartridge is the size of a well plate, and can be used in your automated liquid handler or on a lab bench.
3 Simple Steps
Dispense the dye into the wells on the flip side of the cartridge.
Flip the tab (or have the robot do it)
Check and compare the level of the dye against the graduations
Catalog No. NC2385942
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.