Learn More
Sino Biological Mouse Interleukin-2 / IL-2 Protein 20ug
Supplier:  Sino Biological 51061MNAE20
Encompass_Preferred
A DNA sequence encoding the soluble form of mouse IL2 (P04351) (Ala 21-Gln 169) was expressed, with an initial Met at the C-terminus.
Catalog No. 50-162-2807
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.