Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(ethylcyclopentadienyl)cobalt(II)

Catalog No. 5106451G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-064-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-064-51G Supplier Sigma Aldrich Supplier No. 5106451G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Atomic number of base material: 27 CobaltThis metallocene compound may be encapsulated in single walled carbon nanotubes, resulting in a tailored carbon nano structure of specific electronic functionality.

Specifications

CAS 55940-05-1
Density 1.516 g/mL (at 25°C)
Molecular Formula C14H18Co
Linear Formula C14H18Co
Quantity 1 g
MDL Number MFCD01863655
Synonym (EtCp)2 Co; 1,1 -Diethylcobaltocene
Molecular Weight (g/mol) 245.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.