Learn More
Sino Biological Mouse IL1F8 / IL36b Protein 50ug
Supplier:  Sino Biological 51071MNAE50
Encompass_Preferred
A DNA sequence encoding the mouse IL1F8 (Q9D6Z6) (Met 1-Lys 183) was expressed.
Catalog No. 50-162-2824
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.