Learn More
Sino Biological Mouse IL-21R / Il21R Protein (ECD, His Tag) 100ug
Supplier:  Sino Biological 51184M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Il21r (AAL82636.1) (Met1-Pro236) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.