Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-3-Methylimidazolium Ethyl Sulfate

Catalog No. 516821KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-682-1KGF 1 kg
51-682-100GF 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 51-682-1KGF Supplier Sigma Aldrich Supplier No. 516821KGF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-3-methylimidazolium ethyl sulfate is a water-miscible, aprotic room temperature ionic liquid (RTIL). It can be prepared by reacting 1-methylimidazole with diethyl sulfate.[EMIM][ESO4] has been used to study its effect:As a solvent on the thermal stability of lysozyme.As an additive on potassium hydroxide (KOH) electrolyte properties and hydrogen evolution reaction (HER) reaction kinetics.

Specifications

CAS 342573-75-5
Molecular Formula C8H16N2O4S
Linear Formula C8H16N2O4S
Quantity 1 kg
MDL Number MFCD06798189
Synonym [EMIM][ESO4]
Molecular Weight (g/mol) 236.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.