Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(III) acetylacetonate

Catalog No. 51700310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-700-310G 10 g
51-700-350G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 51-700-310G Supplier Sigma Aldrich Supplier No. 51700310G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron(III) acetylacetonate

Specifications

CAS 14024-18-1
Melting Point 180°C to 182°C
Density 5.24 g/mL (at 25°C)
Molecular Formula C15H21FeO6
Linear Formula Fe(C5H7O2)3
Quantity 10 g
MDL Number MFCD00000020
Synonym 2,4-Pentanedione iron(III) derivative; Fe(acac)3; Ferric acetylacetonate; Iron(III) 2,4-pentanedionate
Molecular Weight (g/mol) 353.17
Percent Purity ≥99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.