Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloroplatinic acid hydrate

Catalog No. 52089625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-089-625G 25 g
52-089-65G 5 g
52-089-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 52-089-625G Supplier Sigma Aldrich Supplier No. 52089625G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloroplatinic acid hydrate

Specifications

Melting Point 60°C
CAS 26023-84-7
Quantity 25 g
Molecular Formula H2Cl6Pt -+ xH2O
Linear Formula H2PtCl6 -+ xH2O
MDL Number MFCD00149909
Synonym Hexachloroplatinic(IV) acid hydrate; Hydrogen hexachloroplatinate(IV) hydrate; Platinic chloride hydrate
Molecular Weight (g/mol) 409.81
Percent Purity ≥99.9% trace metals basis
Density 2.43 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.