Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 53BP1 Recombinant Rabbit Monoclonal Antibody (7H14L12), Alexa Fluor™ Plus 647

Catalog No. 703580RP647
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-358-0RP647 50 μL
50-312-6258 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-358-0RP647 Supplier Invitrogen™ Supplier No. 703580RP647
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Applications Tested: This 53BP1 antibody has been tested by immunocytochemistry and flow cytometric analysis of HeLa cells. This may be used for immunocytochemistry at 10 μg/mL and for flow cytometry at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation:...

53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 53BP1
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7H14L12
Concentration 1.0 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8
Gene TP53BP1
Gene Accession No. Q12888
Gene Alias 53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen Protein corresponding to human 53BP1 [aa241-aa440].
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7158
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.