Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium beta-hydroxypyruvate hydrate

Catalog No. 549135GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-913-5GF 5 g
54-913-1GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-913-5GF Supplier Sigma Aldrich Supplier No. 549135GF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Substrate for transketolase; Preparation of optically pure L-2-hydroxy aldehydes

Specifications

CAS 3369-79-7
Molecular Formula C3H3LiO4 -+ xH2O
Linear Formula HOCH2COCOOLi -+ aq
MDL Number MFCD10567214
Quantity 5 g
Synonym beta-Hydroxypyruvic acid lithium salt hydrate; 3-Hydroxy-2-oxopropanoic acid lithium salt; 3-Hydroxy-2-oxopropionic acid lithium salt
Molecular Weight (g/mol) 109.99 (anhydrous basis)
Percent Purity ≥97.0% (calc. based on dry substance, NT)
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.