Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 55 cal Arc Flash Kit with Voltage Gloves (11)
SDP

Catalog No. 170011774 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1774 Medium
17-001-1769 Small
17-001-1779 Large
17-001-1784 2X-Large
17-001-1789 3X-Large
17-001-1794 4X-Large
17-001-1802 5X-Large
17-001-1803 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1774 Supplier National Safety Apparel™ Supplier No. KIT4SC55MD11
Add to Cart
Add to Cart

Kit includes: 55 cal TECGEN CC short coat and bib overall, Arc flash hood with PureView™ faceshield and hard hat, safety glasses, hood bag and gear bag

  • 30% lighter than standard 40 cal kits
  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Hook and loop at side of bib for easy access

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Gender Unisex
Product Type Arc Flash Kit
Arc Flash Rating 55 cal/cm²
Closure Type FR Hook and Loop
Material Double-Layer TECGEN CC™
Suspender Adjustable
Color Navy
Ankle Style Open
Seam Type Stitched
Size Medium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.