Learn More
Pharmacal Research Laboratories 55 Gallon Drum Ph control Acid Neutralizer
SDP
Supplier:  Pharmacal Research Laboratories 06255F
Encompass
pH Control® is specifically formulated for use where neutralization of acid materials is required prior to disposal. This product is also used as an intermediate compound between an acid and alkaline so that cleaning can be done in the same wash solution. This compound is stable in wash temperatures in excess of 180° F and will not break down under these conditions.
Catalog No. NC9777587
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.